Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR RotafloR dual plug1EA
Catalog No. 502905157
Encompass
Automatic-zero dual stopcock helps eliminate titration errors. Straight-bore with white enamel marks 3mm bore greaseless stopcocks.
Catalog No. 50-290-5157 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS2110501EA
Automatic-zero dual stopcock helps eliminate titration errors. Straight-bore with white enamel marks 3mm bore greaseless stopcocks.