Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR round bottom boilin1EA
Catalog No. 502905477
Encompass
These round bottom boiling flasks have short ring necks and are designed to be used with rubber stoppers. They are precisely tooled for uniform stopper fit. The reinforced rim increases mechanical strength. These flasks are used in applications requiring rigid stoppering and as wash bottles in routine lab washing. Wall thicknesses are carefully controlled to provide the proper balance between thermal expansion and mechanical strength for greater shock resistance. Their excellent chemical durability results in contamination-free results and long life under the toughest lab conditions.
Catalog No. 50-290-5477 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS426012L1EA
These round bottom boiling flasks have short ring necks and are designed to be used with rubber stoppers. They are precisely tooled for uniform stopper fit. The reinforced rim increases mechanical strength. These flasks are used in applications requiring rigid stoppering and as wash bottles in routine lab washing. Wall thicknesses are carefully controlled to provide the proper balance between thermal expansion and mechanical strength for greater shock resistance. Their excellent chemical durability results in contamination-free results and long life under the toughest lab conditions.