Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR serological pipette
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS707810N720EA
Encompass
These sterile disposable Pyrex serological pipettes are designed for the larger-volume user. They are calibrated to deliver (TD) with blow-out and are packed to permit easy access to several pipettes at a time without contaminating the entire case. Similar to No. 7077 except multi-packed in plastic bags.
Catalog No. 50-290-8848