Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR shaker flask with e
Catalog No. 502905719
Encompass
Delong style neck with three extra-deep baffles to enhance gas transfer when used with rotary or reciprocating shakers. Long neck reduces splashing and is designed for two-position polypropylene closures. Closures are sold separately.
Catalog No. 50-290-5719 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS44462L3EA
Delong style neck with three extra-deep baffles to enhance gas transfer when used with rotary or reciprocating shakers. Long neck reduces splashing and is designed for two-position polypropylene closures. Closures are sold separately.