Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR Shorty serological
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS7077B10400EA
Encompass
These sterile disposable shorty Pyrex serological pipettes are calibrated to deliver (TD) with blow-out. They are ideal for use in confined areas such as hoods. The 5 10 and 25 mL sizes have tooled tops and the 50 mL has a polypropylene top end. Negative graduations on each pipet make it applicable for both serological and measuring procedures.
Catalog No. 50-290-8862