Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR Shorty serological
Catalog No. 502908706
Encompass
These sterile disposable shorty Pyrex serological pipettes are calibrated to deliver (TD) with blow-out. They are ideal for use in confined areas such as hoods. The 5 10 and 25 mL sizes have tooled tops and the 50 mL has a polypropylene top end. Negative graduations on each pipet make it applicable for both serological and measuring procedures. Packaged in plastic bags so several can be removed without contaminating the entire pack.
Catalog No. 50-290-8706 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS7078B5400EA
These sterile disposable shorty Pyrex serological pipettes are calibrated to deliver (TD) with blow-out. They are ideal for use in confined areas such as hoods. The 5 10 and 25 mL sizes have tooled tops and the 50 mL has a polypropylene top end. Negative graduations on each pipet make it applicable for both serological and measuring procedures. Packaged in plastic bags so several can be removed without contaminating the entire pack.