Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR single metric scale1EA
Supplier: Sigma Aldrich Fine Chemicals Biosciences CLS30245001EA
Removable PE bumperguard supplied with 25 mL through 1 000 mL sizesThe assortment pack is designed for the low volume user and contains one each of 10 mL 25 mL 50 mL 100 mL 250 mL.