Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR solid glass beads 31L
Catalog No. 502905908
Encompass
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.
Catalog No. 50-290-5908 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS726831LB
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.