Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR solid glass beads 41L
Catalog No. 502905807
Encompass
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.
Catalog No. 50-290-5807 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS726841LB
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.