Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR solid glass beads 5
Catalog No. 502909013
Encompass
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.
Catalog No. 50-290-9013 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS726854LB
These PYREX(R) beads are useful as boiling stones mixing beads and as packing for distillation columns. Beads are packaged in 0.45kg. (1 lb.) bottles (each bottle is one pack) which have a packing volume of approximately 360cm3 or 22 cubic inches.