Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR solution bottle cap1EA
Catalog No. 502908514
Encompass
These Pyrex bottles are designed for storage of SOLUTIONons and are ideal for media preparation. Necks are tooled to increase mechanical strength and for uniform stopper fit.
Catalog No. 50-290-8514 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS1595121EA
These Pyrex bottles are designed for storage of SOLUTIONons and are ideal for media preparation. Necks are tooled to increase mechanical strength and for uniform stopper fit.