Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR square media storag1L
Catalog No. 502905002
Encompass
These heavy duty 1L square bottles are manufactured from PYREX borosilicate glass for chemical and thermal resistance. Square bottles are easier to handle require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing sampling and storage. features include white enamel graduations large marking spot and a glass ridge molded into the bottle to indicate full capacity. Bottles include a linerless one-piece autoclavable orange GL45 threaded polypropylene plug seal cap and a drip-free pouring ring. -Caution Square bottles should NOT be used with bottle top filter units or in other applications involving vacuum pressure as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. -A wide range of optional caps are available for these bottles -Polypropylene colored caps for ease of sorting and identification. -Vented membrane cap with 0.22 mum PTfE hydrophobic membrane. -PBT (polybutylene terepht
Catalog No. 50-290-5002 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS13961L10EA
These heavy duty 1L square bottles are manufactured from PYREX borosilicate glass for chemical and thermal resistance. Square bottles are easier to handle require less space (13-20%) on the shelf or in the autoclave and are ideal for mixing sampling and storage. features include white enamel graduations large marking spot and a glass ridge molded into the bottle to indicate full capacity. Bottles include a linerless one-piece autoclavable orange GL45 threaded polypropylene plug seal cap and a drip-free pouring ring. -Caution Square bottles should NOT be used with bottle top filter units or in other applications involving vacuum pressure as breakage may occur. DO NOT tighten caps immediately after autoclaving as the vacuum resulting from cooling can cause breakage. -A wide range of optional caps are available for these bottles -Polypropylene colored caps for ease of sorting and identification. -Vented membrane cap with 0.22 mum PTfE hydrophobic membrane. -PBT (polybutylene terepht