Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR tissue grinder capa1EA
Catalog No. 502900884
Encompass
Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.
Catalog No. 50-290-0884 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS772271EA
Supplied with two pestles to ensure dissociation of cells into fine particles with minimal damage to cell nuclei. Use the large clearance pestle for initial reduction of soft tissue. Complete the homogenization with the small clearance pestle. Particularly useful in enzyme studies where heat build-up must be avoided.