Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR VISTATM culture tub
Supplier:  Sigma Aldrich Fine Chemicals Biosciences CLS7082516XX200EA
Encompass
These Pyrex Vista culture tubes are supplied with deep-form phenolic caps to facilitate handling and sealing after autoclaving. The blue enamel marking spot gives an excellent surface for pencil notations. Pencil marks are easily erased.
Catalog No. 50-290-5582