Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR VISTATM round-botto
Catalog No. 502900865
Encompass
These round bottom boiling flasks have full length outer joints but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.
Catalog No. 50-290-0865 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS703202504EA
Add to Cart
Add to Cart
These round bottom boiling flasks have full length outer joints but with shorter necks. Wall thicknesses are controlled to provide balance between thermal expansion and mechanical strength for greater shock resistance.