Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR volumetric flask cl1L
Catalog No. 502905937
Encompass
Machine-blown body sealed to heavy-bead heavy-tubing neck for strength 1 and 2 mL sizes are test tube shaped.
Catalog No. 50-290-5937 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS56401L6EA
Machine-blown body sealed to heavy-bead heavy-tubing neck for strength 1 and 2 mL sizes are test tube shaped.