Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR volumetric flask wi
Supplier: Sigma Aldrich Fine Chemicals Biosciences CLS564410012EA
The strength of the flasks have been significantly increased through machine-blown bodies which are sealed to heavy-tubing necks.