Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR wide mouth graduate
Catalog No. 502905720
Encompass
features a GL 45 size PP plug-seal screw-cap on a wide 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage or as a culture flask when optional membrane cap is used.
Catalog No. 50-290-5720 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS49955006EA
features a GL 45 size PP plug-seal screw-cap on a wide 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage or as a culture flask when optional membrane cap is used.