Learn More
Sigma Aldrich Fine Chemicals Biosciences PyrexR wide-mouth graduate1EA
Catalog No. 502900387
Encompass
features a GL 45 size PP plug-seal screw-cap on a wide 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage or as a culture flask when optional membrane cap is used.
Catalog No. 50-290-0387 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CLS49952L1EA
features a GL 45 size PP plug-seal screw-cap on a wide 29 mm neck opening that accommodates larger volume serological pipettes. Also good for media prep and storage or as a culture flask when optional membrane cap is used.