Learn More
Cayman Chemical Pyribencarb, Purity: ≥95%, CAS: 799247-52-2, 100 mg
SDP
Catalog No. 502978444
Encompass_Preferred
A carbamate fungicide; inhibits the production of aflatoxins in A. parasiticus (IC50 = 0.43 µM); inhibits wild-type B. cinerea and strobilurin-resistant B. cinerea mitochondrial complex II (IC50s = 0.079 and 5.1 µM, respectively) and wild-type B. cinerea mitochondrial complex III (IC50 = 0.023 µM); prevents and reduces B. cinerea lesion development on cucumber seedlings at 3 µg/ml; inhibits B. cinerea mycelial growth (EC50 = 0.006 µg/ml)
Catalog No. 50-297-8444 Supplier Cayman Chemical Supplier No. 42362100 MG
A carbamate fungicide; inhibits the production of aflatoxins in A. parasiticus (IC50 = 0.43 µM); inhibits wild-type B. cinerea and strobilurin-resistant B. cinerea mitochondrial complex II (IC50s = 0.079 and 5.1 µM, respectively) and wild-type B. cinerea mitochondrial complex III (IC50 = 0.023 µM); prevents and reduces B. cinerea lesion development on cucumber seedlings at 3 µg/ml; inhibits B. cinerea mycelial growth (EC50 = 0.006 µg/ml)