Learn More
Medchemexpress LLC Pyribencarb | 799247-52-2 | MFCD22200855 | ≥95.0% | 361.83 | C18H20ClN3O3 | 50 MG
SDP
Catalog No. 5000300481
Encompass_Preferred
Pyribencarb is a benzylcarbamate-class fungicide supplied as a research-grade standard. It acts as a Qo inhibitor of cytochrome b (complex III) in the mitochondrial electron transport chain and shows activity against a broad range of plant pathogenic fungi. The compound is intended for analytical, in vitro antifungal, and mode-of-action studies.
  • Inhibits cytochrome b (complex III) via the Qo site, enabling mechanism-of-action research.
  • Active against common plant pathogens such as Botrytis cinerea and Sclerotinia sclerotiorum.
  • Provided as a solid reference material suitable for analytical assays and standards.
  • Storage stability as powder at low temperatures supports long-term archiving.
  • Applicable to in vitro antifungal testing and comparative potency studies.
Catalog No. 50-003-00481 Supplier Medchemexpress LLC Supplier No. HYW02004350MG
Pyribencarb is a benzylcarbamate-class fungicide supplied as a research-grade standard. It acts as a Qo inhibitor of cytochrome b (complex III) in the mitochondrial electron transport chain and shows activity against a broad range of plant pathogenic fungi. The compound is intended for analytical, in vitro antifungal, and mode-of-action studies.
  • Inhibits cytochrome b (complex III) via the Qo site, enabling mechanism-of-action research.
  • Active against common plant pathogens such as Botrytis cinerea and Sclerotinia sclerotiorum.
  • Provided as a solid reference material suitable for analytical assays and standards.
  • Storage stability as powder at low temperatures supports long-term archiving.
  • Applicable to in vitro antifungal testing and comparative potency studies.