Learn More
Medchemexpress LLC Pyribencarb | 799247-52-2 | MFCD22200855 | ≥95.0% | 361.83 g/mol | C18H20ClN3O3 | 100 MG
SDP
Catalog No. 5000301742
Encompass_Preferred
Pyribencarb (CAS 799247-52-2) is a benzylcarbamate-type fungicide provided as an analytical/research standard. It acts at the Qo site of fungal complex III and is used in studies of plant pathogenic fungi, antifungal screening, and fungal biochemistry.
  • High chemical purity suitable for analytical applications.
  • Useful as a research standard for antifungal activity assays.
  • Stable solid form for reliable storage and handling.
  • Provides reproducible inhibition of fungal respiratory pathways.
  • Compatible with common solvents for formulation and testing.
Catalog No. 50-003-01742 Supplier Medchemexpress LLC Supplier No. HYW020043100MG
Pyribencarb (CAS 799247-52-2) is a benzylcarbamate-type fungicide provided as an analytical/research standard. It acts at the Qo site of fungal complex III and is used in studies of plant pathogenic fungi, antifungal screening, and fungal biochemistry.
  • High chemical purity suitable for analytical applications.
  • Useful as a research standard for antifungal activity assays.
  • Stable solid form for reliable storage and handling.
  • Provides reproducible inhibition of fungal respiratory pathways.
  • Compatible with common solvents for formulation and testing.