Learn More
Medchemexpress LLC Pyribencarb | 799247-52-2 | MFCD22200855 | 98.8% | 361.83 | C18H20ClN3O3 | 10 MG
SDP
Catalog No. 5000301432
Encompass_Preferred
Pyribencarb is a benzylcarbamate-type fungicide supplied as a research-grade reagent. It acts as a Qo inhibitor of cytochrome b and is active against a broad range of plant pathogenic fungi. Offered in solid form and as a DMSO solution for laboratory and analytical applications.
  • High purity (98.8%).
  • Molecular formula C18H20ClN3O3, molecular weight 361.83.
  • Available in multiple pack sizes for small-scale research.
  • Provided as solid and 10 mM solution in DMSO for convenience.
  • Suitable for antifungal research and biochemical assays.
Catalog No. 50-003-01432 Supplier Medchemexpress LLC Supplier No. HYW02004310MG
Pyribencarb is a benzylcarbamate-type fungicide supplied as a research-grade reagent. It acts as a Qo inhibitor of cytochrome b and is active against a broad range of plant pathogenic fungi. Offered in solid form and as a DMSO solution for laboratory and analytical applications.
  • High purity (98.8%).
  • Molecular formula C18H20ClN3O3, molecular weight 361.83.
  • Available in multiple pack sizes for small-scale research.
  • Provided as solid and 10 mM solution in DMSO for convenience.
  • Suitable for antifungal research and biochemical assays.