Learn More
Medchemexpress LLC Pyribencarb | 799247-52-2 | MFCD22200855 | 98.8% | 361.83 | C18H20ClN3O3 | 5 MG
SDP
Catalog No. 5000301964
Encompass_Preferred
Pyribencarb is a benzylcarbamate-type fungicide supplied as a research-grade analytical standard. It functions as a Qo inhibitor of fungal respiration and is used as a reference material in laboratory and environmental analyses. Supplied in small pack sizes, the compound requires cold storage and is intended for research use only.
  • Benzylcarbamate-type fungicide and Qo inhibitor.
  • Intended for use as an analytical reference standard in laboratory and environmental testing.
  • Available in small pack sizes suitable for analytical workflows.
  • Molecular formula C18H20ClN3O3; molecular weight 361.83.
  • Store powder at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-003-01964 Supplier Medchemexpress LLC Supplier No. HYW0200435MG
Pyribencarb is a benzylcarbamate-type fungicide supplied as a research-grade analytical standard. It functions as a Qo inhibitor of fungal respiration and is used as a reference material in laboratory and environmental analyses. Supplied in small pack sizes, the compound requires cold storage and is intended for research use only.
  • Benzylcarbamate-type fungicide and Qo inhibitor.
  • Intended for use as an analytical reference standard in laboratory and environmental testing.
  • Available in small pack sizes suitable for analytical workflows.
  • Molecular formula C18H20ClN3O3; molecular weight 361.83.
  • Store powder at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month).