Learn More
AdipoGen Pyribenzoxim
SDP
Supplier:  AdipoGen CDXP0132M500
Encompass_Preferred
Chemical. CAS 168088-61-7. Formula C32H27N5O8. MW 609.59. Synthetic. Unclassified herbicide. Broad-spectrum post-emergence herbicide used to control grasses and polygonums in rice, wheat and other crops. Its mode of action is via the inhibition of acetolactate synthase ALS acetohydroxyacid synthase AHAS. Compound can be used as analytical reference material.
Catalog No. 50-256-0230