Learn More
AdipoGen Pyributicarb
SDP
Supplier:  AdipoGen CDXP0129M250
Encompass_Preferred
Chemical. CAS 88678-67-5. Formula C18H22N2O2S. MW 330.4. Synthetic. Thiocarbamate herbicide with fungicidal activity. Inhibits 4-hydroxyphenyl-pyruvatedioxygenase 4-HPPD in plants that causes bleaching. And targets squalene-epoxidase in sterol biosynthesis in fungus. Compound can be used as analytical reference material.
Catalog No. 50-256-0234