Learn More
Cayman Chemical PyrIdaben-d13 500ug
SDP
Catalog No. 502918475
Encompass_Preferred
An internal standard for the quantification of pyridaben by GC- or LC-MS
Catalog No. 50-291-8475 Supplier Cayman Chemical Supplier No. 39259500 UG
An internal standard for the quantification of pyridaben by GC- or LC-MS