Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pyridazine | 289-80-5 | 80.09 | 10 G
SDP

Catalog No. 5000460325
Encompass_Preferred

Pyridazine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only and not for patient use.

  • Used as a biochemical reagent, biological material, or organic compound for life science related research
  • Appearance: Liquid (Density: 1.103 g/cm³)
  • Color: Colorless to light yellow
  • Solubility in DMSO: 200 mg/mL (2497.19 mM)
  • Storage conditions: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-60325 Supplier Medchemexpress LLC Supplier No. HYW00749410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pyridazine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. This product is for research use only and not for patient use.

  • Used as a biochemical reagent, biological material, or organic compound for life science related research
  • Appearance: Liquid (Density: 1.103 g/cm³)
  • Color: Colorless to light yellow
  • Solubility in DMSO: 200 mg/mL (2497.19 mM)
  • Storage conditions: -20°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.