Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridazine, 98+%

Catalog No. AC131760050
Change view
Click to view available options
Quantity:
5 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC131760050 5 g Glass bottle
AC131760500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC131760050 Supplier Thermo Scientific Chemicals Supplier No. 131760050
Add to Cart

CAS: 289-80-5 | C4H4N2 | 80.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 289-80-5
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00006463
InChI Key PBMFSQRYOILNGV-UHFFFAOYSA-N
Synonym orthodiazine, 1,2-diazine, o-diazine, 1,2-diazabenzene, pyridazin, 1,2-diazin, oizine, pyridazine, pubchem17789, acmc-209h5x
PubChem CID 9259
ChEBI CHEBI:30954
IUPAC Name pyridazine
SMILES C1=CC=NN=C1

Specifications

Melting Point -8°C
Color Yellow to Brown
Density 1.1030g/mL
Boiling Point 208°C
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.5225 to 1.5245
Quantity 5 g
Beilstein 23,89
Specific Gravity 1.103
Merck Index 15,808
Solubility Information Solubility in water: miscible.,miscible with benzene and dmf,freely soluble in methanol,ethanol and ether,practically insoluble in petroleum ether
Formula Weight 80.09
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Pyridazine, 98+%
Show More Show Less
EINECSNumber 206-025-5
RTECSNumber UR5267000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.