Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Pyridin-2-ylmethyl)phosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. P2526200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2526200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2526200MG Supplier TCI America Supplier No. P2526200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 80241-45-8
Molecular Formula C6H8NO3P
Molecular Weight (g/mol) 173.108
MDL Number MFCD16877725
InChI Key WVWCWWLEFNGDBC-UHFFFAOYSA-N
PubChem CID 12730177
IUPAC Name pyridin-2-ylmethylphosphonic acid
SMILES C1=CC=NC(=C1)CP(=O)(O)O

Specifications

Melting Point 287°C
Color White
Quantity 200 mg
Formula Weight 173.11
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Pyridin-2-ylmethyl)phosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.