Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Pyridine
SCI_ED

Catalog No. S25762A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25762A 100 mL Reagent Grade
S25762 50 mL Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25762A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Color Colorless
Density 0.978g/mL
Quantity 100 mL
Packaging Reagent Grade
Assay Percent Range 100% w/v
Linear Formula C5H5N
Formula Weight 79.1
Grade Reagent
Chemical Name or Material Pyridine
Safety handling Check Alert storage code Red
ShelfLife 2 years
DOTInformation DOT Class 3, II, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.