Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pyridine-2,6-Bis(Carboximidamide) Dihydrochloride

Catalog No. 902047250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-204-7250MG 1 g
90-204-71G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-204-7250MG Supplier Sigma Aldrich Supplier No. 902047250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

This pyridyl carboxamidine ligand has been demonstrated by Daniel Weix's lab to enable nickel-catalyzed cross-coupling of diverse basic nitrogen heterocycles with primary and secondary alkyl halides.New ligands for nickel catalysis from diverse pharmaceutical heterocycle libraries

Specifications

CAS 2105938-35-8
Melting Point >300°C
Molecular Formula C9H7N7
Linear Formula C9H7N7
Quantity 1 g
Synonym Weix 2-pyridyl-N-cyanocarboxamidine ligand for nickel catalysis
Molecular Weight (g/mol) 213.2 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.