Learn More
Medchemexpress LLC Pyridine-2,6-dicarboxylic acid | 499-83-2 | MFCD00006299 | 100.0% | 167.12 g/mol | C7H5NO4 | 10g
SDP
Catalog No. 5000266728
Encompass_Preferred
Pyridine-2 6-dicarboxylic acid is a dicarboxylic acid containing a pyridine ring with unique chemical properties that make it an important ingredient in a variety of industrial processes Pyridine-2 6-dicarboxylic acid is commonly used as a ligand for metal ions a building block in the synthesis of pharmaceuticals and organic compounds and a precursor in the production of pigments and dyes
Catalog No. 50-002-66728 Supplier Medchemexpress LLC Supplier No. HYY102410G
Pyridine-2 6-dicarboxylic acid is a dicarboxylic acid containing a pyridine ring with unique chemical properties that make it an important ingredient in a variety of industrial processes Pyridine-2 6-dicarboxylic acid is commonly used as a ligand for metal ions a building block in the synthesis of pharmaceuticals and organic compounds and a precursor in the production of pigments and dyes