Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-2-carbonyl Chloride Hydrochloride 93.0+%, TCI America™
SDP

Catalog No. P06195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P06195G 5 g
P061925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P06195G Supplier TCI America Supplier No. P06195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 39901-94-5
Molecular Formula C6H5Cl2NO
Molecular Weight (g/mol) 178.01
MDL Number MFCD00060193
InChI Key VIPHVHVAGBKHGR-UHFFFAOYSA-N
Synonym Picolinoyl Chloride Hydrochloride
PubChem CID 135033
IUPAC Name hydrogen pyridine-2-carbonyl chloride chloride
SMILES [H+].[Cl-].ClC(=O)C1=CC=CC=N1

Specifications

Color Red-White
Quantity 5 g
UN Number 3261
Formula Weight 178.01
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyridine-2-carbonyl Chloride Hydrochloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.