Learn More
Medchemexpress LLC Pyridine, 2-methyl-3-[(2S)-2-pyrrolidinylmethoxy]-, hydrochloride (1:2). | 161416-61-1 | 99.1% | 265.18 | 50 MG
SDP
Catalog No. 5000345033
Encompass_Preferred
Pozanicline dihydrochloride is an orally bioavailable nicotinic acetylcholine receptor (nAChR) agonist, highly selective for α4β2- and α6β2-nAChR subtypes. It has been shown to reverse nicotine withdrawal-induced deficits in contextual fear conditioning in mice.
  • Orally bioavailable nicotinic acetylcholine receptor (nAChR) agonist.
  • Highly selective for α4β2- and α6β2-nAChR subtypes.
  • Involved in Phase 2 clinical trials for Attention Deficit Hyperactivity Disorder, Alzheimer's Disease, and Nicotine Addiction.
  • Reverses nicotine withdrawal-induced deficits in contextual fear conditioning in mice.
Catalog No. 50-003-45033 Supplier Medchemexpress LLC Supplier No. HY11016050MG
Pozanicline dihydrochloride is an orally bioavailable nicotinic acetylcholine receptor (nAChR) agonist, highly selective for α4β2- and α6β2-nAChR subtypes. It has been shown to reverse nicotine withdrawal-induced deficits in contextual fear conditioning in mice.
  • Orally bioavailable nicotinic acetylcholine receptor (nAChR) agonist.
  • Highly selective for α4β2- and α6β2-nAChR subtypes.
  • Involved in Phase 2 clinical trials for Attention Deficit Hyperactivity Disorder, Alzheimer's Disease, and Nicotine Addiction.
  • Reverses nicotine withdrawal-induced deficits in contextual fear conditioning in mice.