Learn More
Medchemexpress LLC Pyridine-3,5-dicarboxylic acid | 499-81-0 | 167.12 | 1 KG
SDP
Catalog No. 5000448835
Encompass_Preferred
Pyridine-3,5-dicarboxylic acid is a drug intermediate used to construct new metal-organic framework materials.
  • Store as powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (50 mg/mL with warming).
  • For research use only.
Catalog No. 50-004-48835 Supplier Medchemexpress LLC Supplier No. HYW0020001KG
Pyridine-3,5-dicarboxylic acid is a drug intermediate used to construct new metal-organic framework materials.
  • Store as powder at -20°C for up to 3 years or 4°C for up to 2 years.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • Appears as a white to off-white solid.
  • Soluble in DMSO (50 mg/mL with warming).
  • For research use only.