Learn More
Medchemexpress LLC Pyridine-3,5-dicarboxylic acid | 499-81-0 | 167.12 | 100 G
SDP
Catalog No. 5000446426
Encompass_Preferred
Pyridine-3,5-dicarboxylic acid is a drug intermediate used to construct new metal-organic framework materials.
  • Drug intermediate
  • Can be used to construct new metal-organic framework materials
Catalog No. 50-004-46426 Supplier Medchemexpress LLC Supplier No. HYW002000100G
Pyridine-3,5-dicarboxylic acid is a drug intermediate used to construct new metal-organic framework materials.
  • Drug intermediate
  • Can be used to construct new metal-organic framework materials