Learn More
Medchemexpress LLC Pyridine-3,5-dicarboxylic acid | 499-81-0 | 167.12 | 50 G
SDP
Catalog No. 5000446922
Encompass_Preferred
Pyridine-3,5-dicarboxylic acid is a drug intermediate that can be used to construct new metal-organic framework materials. It is intended for research use only.
  • Used as a drug intermediate.
  • Can be used to construct new metal-organic framework materials.
  • Intended for research use only.
Catalog No. 50-004-46922 Supplier Medchemexpress LLC Supplier No. HYW00200050G
Pyridine-3,5-dicarboxylic acid is a drug intermediate that can be used to construct new metal-organic framework materials. It is intended for research use only.
  • Used as a drug intermediate.
  • Can be used to construct new metal-organic framework materials.
  • Intended for research use only.