Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-3-carboxaldoxime, 98%, Thermo Scientific™

Catalog No. AAB2567922
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2567922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2567922 Supplier Thermo Scientific Chemicals Supplier No. B2567922

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-92-6
Molecular Formula C6H6N2O
Molecular Weight (g/mol) 122.13
MDL Number MFCD00006409,MFCD00006409
InChI Key YBKOPFQCLSPTPV-VMPITWQZSA-N
Synonym nicotinaldoxime, 3-pyridinealdoxime, nicotinaldehyde oxime, pyridine-3-aldoxime, 3-pyridinecarboxaldehyde, oxime, nicotinaldehyde, oxime, 3-pyridine-aldoxime, pyridine, 3-formyl-, oxime, hydroxyimino-3-pyridylmethane, pyridinaldoxim
PubChem CID 5371219
IUPAC Name (NZ)-N-(pyridin-3-ylmethylidene)hydroxylamine
SMILES O\N=C\C1=CC=CN=C1

Specifications

Melting Point 148°C to 151°C
Quantity 100 g
Formula Weight 122.13
Percent Purity 98%
Chemical Name or Material Pyridine-3-carboxaldoxime
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECSNumber UT4060000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.