Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine-3-carboxamidine hydrochloride 0.5 hydrate, 95%, Thermo Scientific™

Catalog No. MO07766DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07766DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07766DE Supplier Thermo Scientific Chemicals Supplier No. MO07766DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 871825-82-0
Molecular Formula C12H18Cl2N6O
Molecular Weight (g/mol) 333.217
InChI Key AOHLMHDFGXVZAQ-UHFFFAOYSA-N
Synonym pyridine-3-carboximidamide hemihydrate hydrochloride
PubChem CID 90477204
IUPAC Name pyridine-3-carboximidamide;hydrate;dihydrochloride
SMILES C1=CC(=CN=C1)C(=N)N.C1=CC(=CN=C1)C(=N)N.O.Cl.Cl

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 166.61
Percent Purity 95%
Chemical Name or Material Pyridine-3-carboxamidine hydrochloride 0.5 hydrate
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.