Learn More
Medchemexpress LLC Pyridine, 3-chloro-2-(2-methylpropoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- | 1387634-81-2 | 311.62 | 5 G
SDP
Catalog No. 5000447096
Encompass_Preferred
Pyridine, 3-chloro-2-(2-methylpropoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active compounds. This product, identified by its CAS number 1387634-81-2 and molecular weight of 311.62, demonstrates a purity of 95.0% by NMR. It requires specific storage conditions to maintain its integrity.
  • Used as a versatile drug intermediate
  • Essential for the synthesis of active pharmaceutical compounds
  • High purity of 95.0% (NMR)
  • Requires sealed storage at -20°C, protected from moisture and light
  • Stable for 6 months at -80°C or 1 month at -20°C when in solvent, under sealed, dry, and dark conditions
Catalog No. 50-004-47096 Supplier Medchemexpress LLC Supplier No. HYW0006185G
Pyridine, 3-chloro-2-(2-methylpropoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)- is a chemical compound primarily utilized as a drug intermediate in the synthesis of various active compounds. This product, identified by its CAS number 1387634-81-2 and molecular weight of 311.62, demonstrates a purity of 95.0% by NMR. It requires specific storage conditions to maintain its integrity.
  • Used as a versatile drug intermediate
  • Essential for the synthesis of active pharmaceutical compounds
  • High purity of 95.0% (NMR)
  • Requires sealed storage at -20°C, protected from moisture and light
  • Stable for 6 months at -80°C or 1 month at -20°C when in solvent, under sealed, dry, and dark conditions