Learn More
Medchemexpress LLC Pyridine, 3-ethynyl- | 2510-23-8 | 100.0% | 103.12 | 10 G
SDP
Catalog No. 5000404979
Encompass_Preferred
3-Ethynylpyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. It is for research use only and not sold to patients.
  • Biochemical reagent for life science research.
  • Click chemistry reagent with an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.
Catalog No. 50-004-04979 Supplier Medchemexpress LLC Supplier No. HY3460410G
3-Ethynylpyridine is a biochemical reagent that can be used as a biological material or organic compound for life science related research. It is a click chemistry reagent, containing an alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. It is for research use only and not sold to patients.
  • Biochemical reagent for life science research.
  • Click chemistry reagent with an alkyne group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups.