Learn More
Medchemexpress LLC Pyridine, 4-[2-(2-fluorophenyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl]- | 1784751-19-4 | 99.9% | 333.33 | C20H13F2N3 | 25 MG
SDP
Catalog No. 5000215286
Encompass_Preferred
TA-02 is a small-molecule p38 MAPK inhibitor used for research applications. It is an analog of SB 203580 and selectively inhibits TGFBR-2, showing potent activity with an IC50 of approximately 20 nM. The compound is provided as a solid and as a 10 mM solution in DMSO and is accompanied by a datasheet and certificate of analysis.
Catalog No. 50-002-15286 Supplier Medchemexpress LLC Supplier No. HY10011525MG
TA-02 is a small-molecule p38 MAPK inhibitor used for research applications. It is an analog of SB 203580 and selectively inhibits TGFBR-2, showing potent activity with an IC50 of approximately 20 nM. The compound is provided as a solid and as a 10 mM solution in DMSO and is accompanied by a datasheet and certificate of analysis.