Learn More
Medchemexpress LLC Pyridine, 4-[2-(2-fluorophenyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl]- | 1784751-19-4 | 99.9% | 333.33 | C20H13F2N3 | 5 MG
SDP
Catalog No. 5000215362
Encompass_Preferred
TA-02 is a small-molecule inhibitor of p38 MAPK used in research to probe MAPK signaling and cardiogenic pathways. It inhibits phosphorylation of downstream proteins such as MAPKAPK2 and HSP27 and shows activity against TGFBR-2.
  • Potent p38 MAPK inhibition (IC50 20 nM).
  • High purity suitable for research applications (99.85%).
  • Molecular formula C20H13F2N3; molecular weight 333.33.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • Useful for studies of cardiogenesis and TGFBR-2 related signaling.
Catalog No. 50-002-15362 Supplier Medchemexpress LLC Supplier No. HY1001155MG
TA-02 is a small-molecule inhibitor of p38 MAPK used in research to probe MAPK signaling and cardiogenic pathways. It inhibits phosphorylation of downstream proteins such as MAPKAPK2 and HSP27 and shows activity against TGFBR-2.
  • Potent p38 MAPK inhibition (IC50 20 nM).
  • High purity suitable for research applications (99.85%).
  • Molecular formula C20H13F2N3; molecular weight 333.33.
  • Available as small solid quantities and as a 10 mM solution in DMSO.
  • Useful for studies of cardiogenesis and TGFBR-2 related signaling.