Learn More
Medchemexpress LLC Pyridine, 5-cyclopropyl-2-[4-(2,6-difluorophenoxy)-5-methyl-3-(1-methylethoxy)-1H-pyrazol-1-yl] | 1800296-63-2 | 98.7% | C21H20F3N3O2 | 10MG
SDP
Catalog No. 5000259344
Encompass_Preferred
DHODH-IN-1 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor, reported as compound 18d, used in research on pyrimidine biosynthesis, antiviral activity, and cell proliferation. It shows strong DHODH inhibition (IC50 ≈ 25 nM) and is supplied as a high-purity research reagent with recommended dissolution in DMSO.
  • Potent DHODH inhibitor with IC50 ≈ 25 nM.
  • High purity (98.65%).
  • Molecular formula C21H20F3N3O2; molecular weight 403.40.
  • Available in small milligram pack sizes suitable for in vitro studies.
  • Recommended solvent: DMSO; in vivo formulation protocol provided.
  • Suitable for antiviral and cell-proliferation research.
  • For research use only.
Catalog No. 50-002-59344 Supplier Medchemexpress LLC Supplier No. HY13528210MG
DHODH-IN-1 is a potent dihydroorotate dehydrogenase (DHODH) inhibitor, reported as compound 18d, used in research on pyrimidine biosynthesis, antiviral activity, and cell proliferation. It shows strong DHODH inhibition (IC50 ≈ 25 nM) and is supplied as a high-purity research reagent with recommended dissolution in DMSO.
  • Potent DHODH inhibitor with IC50 ≈ 25 nM.
  • High purity (98.65%).
  • Molecular formula C21H20F3N3O2; molecular weight 403.40.
  • Available in small milligram pack sizes suitable for in vitro studies.
  • Recommended solvent: DMSO; in vivo formulation protocol provided.
  • Suitable for antiviral and cell-proliferation research.
  • For research use only.