Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, 99.8%, MilliporeSigma™

Catalog No. 50947687 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-687 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-687 Supplier MilliporeSigma Supplier No. PX2012P1
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.10
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH 8.81 (H2O, 20°C)
Melting Point −42°C
Quantity 4 L
Flash Point 17°C
Packaging Glass Bottle (Pre-packed)
Residue after Evaporation 5 ppm max
Assay Percent Range 99.8% min
Vapor Pressure 26.7 hPa (25°C)
Boiling Point 115°C (1013 hPa)
Solubility Information 1000 g/L Soluble
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.