Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, >99%, MP Biomedicals™

Catalog No. ICN15198383 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15198383 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15198383 Supplier MP Biomedicals, Inc Supplier No. 0215198383
Add to Cart
Add to Cart

Pyridine acts as a solvent for anhydrous mineral salts, organic and inorganic compounds.

  • Pyridine is a synthetic intermediate in laboratory and industry.
  • Its weakly basic nature allows it to act as a proton acceptor in acid base reactions, and to form salts with strong acids.
  • In the acylation of alcohols, pyridine can act as a nucleophilic catalyst.
  • Various reports have described the use of pyridine to prepare such compounds as indanones, 2 cyclopentenones, polycarbonates, symmetric aromatic trisulfides and tetrasulfides, and calcium-η 2-pyrazolato complexes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

Color Colorless to Yellow
Density 0.982 (water = 1)
Assay Percent Range >99%
Quantity 250 mL
Formula Weight 79.1
Physical Form Liquid
Chemical Name or Material Pyridine
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.