Learn More
Sigma Aldrich Fine Chemicals Biosciences Pyridine ACS reagent 99.0
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 3605702.5L
Encompass
Pyridine (Pyr) is a nitrogen containing six membered heterocyclic compound commonly used as a base. It is a colorless liquid with a fishy odor. Pyridine moiety is present as a major component in a number of compounds like pyridine nucleotides pyridine alkaloids pyridine-based polymers and pyridine-containing macrocycles. It has wide application as a solvent catalyst and as an intermediate for synthesis. IR Raman and UV spectra of pyridine has been analyzed and the values obtained were used to calculate its thermodynamic properties. Photodegradation and bacterial degradation of Pyr has been studied.
Catalog No. 50-289-8155