Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyridine, ACS Reagent, ≥99%, Solstice

Catalog No. 60046626 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-626 2.5 L Glass Bottle
60-046-624 100 mL Glass Bottle
60-046-627 500 mL Glass Bottle
60-046-625 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-626 Supplier Solstice Supplier No. 3605702.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-86-1
Molecular Formula C5H5N
Molecular Weight (g/mol) 79.102
MDL Number MFCD00011732
InChI Key JUJWROOIHBZHMG-UHFFFAOYSA-N
Synonym azabenzene, azine, pyridin, piridina, pirydyna, pyridin german, pirydyna polish, piridina italian, rcra waste number u196, caswell no. 717
PubChem CID 1049
ChEBI CHEBI:16227
IUPAC Name pyridine
SMILES C1=CC=NC=C1

Specifications

pH 8.8 (at 20°C)
Melting Point -42°C
Color Colorless
Density 0.978g/mL at 25°C
Quantity 2.5 L
Explosion Limit 1.8% to 12.4% (V)
Flash Point 17°C (62.6°F)
Ignition Point 899°F
Odor Unpleasant
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 10mmHg (13.2°C), 20mmHg (25°C)
UN Number UN1282
Beilstein 103233
Boiling Point 115°C
Solubility Information Completely miscible in water
Formula Weight 79.1g/mol
Percent Purity ≥99%
Chemical Name or Material Pyridine
Show More Show Less
Hazard Statement P210-P280-P305 + P351 + P338
DOTInformation class 3 - PG 2 - Pyridine
RTECSNumber 203-809-9
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.