Learn More
Pyridine, for GC and non-aqueous titrations, >99.8%, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| CAS Max % | 100.0000% |
| Density | 0.983 g/cm3 |
| Quantity | 4 x 4 L |
| Flash Point | 20°C |
| Packaging | Glass Bottle |
| UN Number | UN1282 |
| Beilstein | 103233 |
| Boiling Point | 115.25°C |
| Formula Weight | 79.1 |
| Percent Purity | >99.8% |
| Show More |
Safety and Handling
| ShelfLife | 547 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Pyridine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.